N,N'-Diphenyl-N,N'-bis(p-tolyl)-1,4-phenylenediamine ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:138171-14-9
:
Formula:C₃₂H₂₈N₂
MDL Number:MFCD01310665
:440.59 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:191.0 to 195.0 deg-C
Specification Test Results
Frequently Bought Together