Order Entry
Netherlands
ContactUsLinkComponent
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
Catalog # TCIAD4889-100MG
Supplier:  TCI
CAS Number:  
undefined
5,5'''-Didodecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥96.0% (by HPLC)
Catalog # TCIAD4889-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Elemental analysis (Sulfuride):
    18.20 to 20.20 %

Specification Test Results

About this item

 :
Melting Pt:166°C
 :667.14 g/mol
CAS Number:153561-79-6
Formula:C₄₀H₅₈S₄