
(4,4'-Di-tert-butyl-2,2'-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosphate ≥85.0% (by HPLC)
Catalog # TCIAD4887-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 85.0 area%
- Elemental analysis (Nitrogen):5.20 to 6.50 %
Specification Test Results
About this item
:
:913.97 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:676525-77-2
Formula:C₄₀H₄₀F₆IrN₄P
MDL Number:MFCD27952560