(R)-(+)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:179866-74-1
:
Formula:C₂₄H₂₀Br₂O₄
MDL Number:MFCD03788936
:532.23 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:133.0 to 137.0 deg-C
- Specific rotation [a]20/D:+20.0 to +23.0 deg (C=1,CHCl3)
Specification Test Results
Frequently Bought Together