Order Entry
Netherlands
ContactUsLinkComponent
2,4-Dichlorothieno[3,2-d]pyrimidine ≥98.0% (by GC)
2,4-Dichlorothieno[3,2-d]pyrimidine ≥98.0% (by GC)
Catalog # TCIAD4864-1G
Supplier:  TCI
CAS Number:  
2,4-Dichlorothieno[3,2-d]pyrimidine ≥98.0% (by GC)
Catalog # TCIAD4864-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    136.0 to 140.0 deg-C

Specification Test Results

About this item

 :
 :205.0645 g/mol
CAS Number:16234-14-3
Formula:C₆H₂Cl₂N₂S
MDL Number:MFCD08448158