N,N'-Diphenyl-N,N'-bis[4'-(diphenylamino)biphenyl-4-yl]benzidine ≥98.0% (by HPLC, total nitrogen)
: TCI
:167218-46-4
:
:C₇₂H₅₄N₄
:MFCD03093703
:975.25 g/mol
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:239.0 to 243.0 deg-C
Frequently Bought Together