4,6-Dibromodibenzothiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:669773-34-6
:
Formula:C₁₂H₆Br₂S
MDL Number:MFCD18451473
:342.048 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:184.0 to 188.0 deg-C
Specification Test Results
Frequently Bought Together