Order Entry
Netherlands
ContactUsLinkComponent
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole ≥98.0% (by HPLC, titration analysis)
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4808-200MG
Supplier:  TCI
   Safer Human and Environmental Health
CAS Number:  
(2S,3R)-3,4-Dihydro-3-isopropyl-2-phenyl-2H-pyrimido[2,1-b]benzothiazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4808-200MG
Supplier:  TCI
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    197.0 to 201.0 deg-C
  • Specific rotation [a]20/D:
    343.0 to 353.0 deg (C=0.5,CHCL3)

Specification Test Results

About this item

 :
 :308.44 g/mol
CAS Number:1203507-02-1
Formula:C₁₉H₂₀N₂S
MDL Number:MFCD28411847