Order Entry
Netherlands
ContactUsLinkComponent
3,7-Dimethyl-2,6-octadienyl butyrate ≥95.0% (by GC)
Irritant
3,7-Dimethyl-2,6-octadienyl butyrate ≥95.0% (by GC)
Catalog # TCIAD4796-25ML
Supplier:  TCI
CAS Number:  
Irritant
3,7-Dimethyl-2,6-octadienyl butyrate ≥95.0% (by GC)
Catalog # TCIAD4796-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

Specification Test Results

About this item

 :
 :224.34 g/mol
 :GHS07
CAS Number:2173-82-2
Formula:C₁₄H₂₄O₂