
Di-1-adamantylphosphine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:131211-27-3
:
MDL Number:MFCD06658117
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:176.0 to 180.0 deg-C
Specification Test Results
Frequently Bought Together