Order Entry
Netherlands
ContactUsLinkComponent
 
4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAD4734-200MG
Supplier:  TCI
CAS Number:  
4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAD4734-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    153.0 to 157.0 deg-C

Specification Test Results

About this item

 :
 :329.94 g/mol
CAS Number:1295502-53-2
Formula:C₆Br₂F₂N₂S
MDL Number:MFCD28369502