
Dimethoxy(diisobutyl)silane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17980-32-4
:
Formula:C₁₀H₂₄O₂Si
MDL Number:MFCD00236510
Boiling Pt:72°C
Flash Pt:102°C
:204.38 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together