Order Entry
Netherlands
ContactUsLinkComponent
6,7-Dimethoxy-4-hydroxyquinoline ≥98.0% (by HPLC, titration analysis)
Irritant
6,7-Dimethoxy-4-hydroxyquinoline ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
6,7-Dimethoxy-4-hydroxyquinoline ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    227.0 to 231.0 deg-C

Specification Test Results

Frequently Bought Together