
3,6-Dibromophenanthrene-9,10-dione ≥95.0% (by HPLC)
Catalog # TCIAD4692-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g/mol
- Seq:9220
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:289.0 to 293.0 deg-C
Specification Test Results
About this item
:
:366.01 g/mol
:GHS07
CAS Number:53348-05-3
Formula:C₁₄H₆Br₂O₂
MDL Number:MFCD00451676