
2,6-Dichloro-4-(trifluoromethyl)pyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:39890-98-7
:
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD00042246
:215.99 g/mol
Flash Pt:85°C
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together