
3,5-Dibromophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:117695-55-3
:
Formula:C₆H₅BBr₂O₂
MDL Number:MFCD01075725
:279.72 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 107.0 %
Specification Test Results
Related Information
Frequently Bought Together