
5,8-Dibromo-2,3-bis(4-methoxyphenyl)quinoxaline ≥98.0% (by HPLC)
TCIAD4653-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):5.30 to 5.90 %
:
:500.19 g/mol
:GHS07
:162967-90-0
:C₂₂H₁₆Br₂N₂O₂