2,6-Dibromo-9,10-di(2-naphthyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:561064-15-1
:
Formula:C₃₄H₂₀Br₂
MDL Number:MFCD16660916
:588.34 g/mol
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together