Order Entry
Netherlands
ContactUsLinkComponent
9,9-Di(p-tolyl)fluorene-2-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Irritant
9,9-Di(p-tolyl)fluorene-2-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Catalog # TCIAD4642-200MG
Supplier:  TCI
CAS Number:  
Irritant
9,9-Di(p-tolyl)fluorene-2-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Catalog # TCIAD4642-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    97.0 to 105.0 %

Specification Test Results

About this item

 :
 :390.29 g/mol
 :GHS07
CAS Number:1193104-83-4
Formula:C₂₇H₂₃BO₂
MDL Number:MFCD23115730