Order Entry
Netherlands
ContactUsLinkComponent
1,5-Dimethyl-1H-pyrazole ≥98.0% (by titrimetric analysis)
Flammable
Irritant
1,5-Dimethyl-1H-pyrazole ≥98.0% (by titrimetric analysis)
Catalog # TCIAD4630-5ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
1,5-Dimethyl-1H-pyrazole ≥98.0% (by titrimetric analysis)
Catalog # TCIAD4630-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :96.1300 g/mol
Boiling Pt:158°C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:694-31-5
Formula:C₅H₈N₂
MDL Number:MFCD01646129