
2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:89402-42-6
:
Formula:C₆H₂F₅N
MDL Number:MFCD04972837
:183.08 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together