2,8-Dimethylanthra[2,3-b:7,6-b']dithiophene, purified by sublimation
Catalog # TCIAD4618-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Elemental analysis (Carbon):71.50 to 79.00 %
- Elemental analysis (Sulfuride):19.00 to 21.00 %
Specification Test Results
About this item
:
:318.45 g/mol
CAS Number:1392416-39-5
Formula:C₂₀H₁₄S₂