
2,4-Dihydroxy-5-methoxypyrimidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6623-81-0
:
Formula:C₅H₆N₂O₃
MDL Number:MFCD00187903
:142.1100 g/mol
Melting Pt:344°C
EINECS:229-580-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together