Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dibromo-4,8-bis[(2-butyl-n-octyl)oxy]benzo[1,2-b:4,5-b']dithiophene ≥94.0% (by HPLC)
2,6-Dibromo-4,8-bis[(2-butyl-n-octyl)oxy]benzo[1,2-b:4,5-b']dithiophene ≥94.0% (by HPLC)
Catalog # TCIAD4592-200MG
Supplier:  TCI
CAS Number:  
2,6-Dibromo-4,8-bis[(2-butyl-n-octyl)oxy]benzo[1,2-b:4,5-b']dithiophene ≥94.0% (by HPLC)
Catalog # TCIAD4592-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (HPLC):
    min. 94.0 area%

Specification Test Results

About this item

 :
 :716.72 g/mol
CAS Number:1336893-15-2
Formula:C₃₄H₅₂Br₂O₂S₂