
2,5-Dibromothiophene-3-carboxylic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7311-70-8
:
Formula:C₅H₂Br₂O₂S
MDL Number:MFCD08445656
:285.94 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together