Diethylene Glycol bis(2-propynyl) ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:126422-57-9
:
Formula:C₁₀H₁₄O₃
MDL Number:MFCD22201523
:182.22 g/mol
Boiling Pt:103°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together