
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:42389-54-8
:
Formula:C₁₀H₂₂N₂
MDL Number:MFCD06658428
:170.30 g/mol
Boiling Pt:111°C
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together