
2,6-Dibromobenzenecarbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:67713-23-9
:
Formula:C₇H₄Br₂O
MDL Number:MFCD11040410
:263.93 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:90.0 to 94.0 deg-C
Specification Test Results
Frequently Bought Together