
5,6-Diaminobenzimidazolinone ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:55621-49-3
:
Formula:C₇H₈N₄O
MDL Number:MFCD00994340
:164.17 g/mol
EINECS:259-728-4
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together