Order Entry
Netherlands
ContactUsLinkComponent
4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole ≥98.0% (by GC)
4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAD4529-200MG
Supplier:  TCI
CAS Number:  
4,7-Dibromo-5,6-di-n-octyloxy-2,1,3-benzothiadiazole ≥98.0% (by GC)
Catalog # TCIAD4529-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    43.0 to 47.0 deg-C

Specification Test Results

About this item

 :
 :550.39 g/mol
CAS Number:1192352-08-1
 :MFCD19440967
Formula:C₂₂H₃₄Br₂N₂O₂S
MDL Number:MFCD19940667