2,6-Dibromo-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene ≥97.0% (by HPLC)
Catalog # TCIAD4524-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:604.50 g/mol
CAS Number:1226782-13-3
Formula:C₂₆H₃₆Br₂O₂S₂
MDL Number:MFCD17018552