
7-(Diethylamino)coumarin-3-carbonitrile ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:51473-74-6
:
Formula:C₁₄H₁₄N₂O₂
MDL Number:MFCD00068688
:242.28 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:228.0 to 232.0 deg-C
Specification Test Results
Frequently Bought Together