Dipotassium-(R)-1,1'-binaphthyl-2,2'-disulphonate ≥98.0% (by HPLC)
TCIAD4445-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Ion exchange titration):min. 95.0 %
- Specific rotation [a]20/D:+60.0 to +65.0 deg (C=1, MeOH)
:
:490.63 g/mol
:1092934-19-4
:C₂₀H₁₄O₆S₂