
5,5'-Dimethyl-2,2'-bipiperidine (mixture of isomers) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1378805-84-5
:
Formula:C₁₂H₂₄N₂
:196.34 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together