D 102 ≥96.0% (by HPLC)
Catalog # TCIAD4430-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to Amber to Dark red powder to crystal
- Purity (HPLC):min. 96.0 area%
Specification Test Results
About this item
:
:614.78 g/mol
CAS Number:652145-28-3
Formula:C₃₇H₃₀N₂O₃S₂