Order Entry
Netherlands
ContactUsLinkComponent
4-(N,N-Dimethylamino)phenylboronic acid (contains varying amounts of Anhydride)
Irritant
4-(N,N-Dimethylamino)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI
Certificates
Irritant
4-(N,N-Dimethylamino)phenylboronic acid (contains varying amounts of Anhydride)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    97.0 to 113.0 %

Specification Test Results

Related Information

Frequently Bought Together