
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:65355-08-0
:
Formula:C₂₀H₂₀Br₂O₂
MDL Number:MFCD03093977
:452.1800 g/mol
Melting Pt:149°C
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
Specification Test Results
Frequently Bought Together