Dimethyl-1,4-naphthalenedicarboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7487-15-2
:
Formula:C₁₄H₁₂O₄
MDL Number:MFCD04038075
:244.25 g/mol
Boiling Pt:161°C
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 69.0 deg-C
Specification Test Results
Frequently Bought Together