Dimethyl decahydro-1,4-naphthalenedicarboxylate (mixture of isomers) ≥97.0% (by GC)
Catalog # TCIAD4399-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 % (total of isomers)
Specification Test Results
About this item
:
:254.33 g/mol
Boiling Pt:178°C
CAS Number:3068-03-9
Formula:C₁₄H₂₂O₄