Dipyrido[3,2-a:2',3'-c]phenazine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:19535-47-8
:
Formula:C₁₈H₁₀N₄
MDL Number:MFCD04113602
:282.31 g/mol
Melting Pt:248°C
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together