
2,5-Dibromothiophene 1,1-dioxide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4376-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:273.93 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:89088-95-9
Formula:C₄H₂Br₂O₂S