(S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4347-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:129.0 to 133.0 deg-C
- Specific rotation [a]20/D:+47.0 to +51.0 deg (C=1,1mol/L HCl)
Specification Test Results
About this item
:
:232.3214 g/mol
CAS Number:27262-40-4
Formula:C₁₄H₂₀N₂O
MDL Number:MFCD09038786