Order Entry
Netherlands
ContactUsLinkComponent
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by GC)
Irritant
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by GC)
Catalog # TCIAD4341-1G
Supplier:  TCI
CAS Number:  
Irritant
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) ≥98.0% (by GC)
Catalog # TCIAD4341-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :262.11 g/mol
Melting Pt:257°C
 :GHS07
CAS Number:121207-31-6
Formula:C₁₄H₁₇BF₂N₂
MDL Number:MFCD00467372