Bestelling invoeren
Netherlands
ContactUsLinkComponent
4,6-Dimethylpyrimidine ≥98.0% (by GC, titration analysis)
Flammable
Irritant
4,6-Dimethylpyrimidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Flammable
Irritant
4,6-Dimethylpyrimidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together