Order Entry
Netherlands
ContactUsLinkComponent
2,2'-Diamino-4,4'-bithiazole ≥98.0% (by HPLC, titration analysis)
Irritant
2,2'-Diamino-4,4'-bithiazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4273-200MG
Supplier:  TCI
CAS Number:  
Irritant
2,2'-Diamino-4,4'-bithiazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4273-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :198.26 g/mol
 :GHS07
CAS Number:58139-59-6
Formula:C₆H₆N₄S₂
MDL Number:MFCD01740587