
7-(Diethylamino)coumarin-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Catalog # TCIAD4238-100MG
Leverancier: TCI
CAS-nummer:
Specificaties
- Pk:100 mg
- Seq:2440
Specificaties
Specificatie testresultaten
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:227.0 to 231.0 deg-C
Specificatie testresultaten
Over dit artikel
:
:261.28 g/mol
:GHS07
CAS-nummer:50995-74-9
Formule:C₁₄H₁₅NO₄
MDL-nummer:MFCD00068721