
2,5-Diamino-4,6-dichloropyrimidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:55583-59-0
:
Formula:C₄H₄Cl₂N₄
MDL Number:MFCD00792652
:179.0100 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together