Order Entry
Netherlands
ContactUsLinkComponent
Daptomycin ≥94.0% (by HPLC, total nitrogen)
Daptomycin ≥94.0% (by HPLC, total nitrogen)
Catalog # TCIAD4229-100MG
Supplier:  TCI
CAS Number:  
Daptomycin ≥94.0% (by HPLC, total nitrogen)
Catalog # TCIAD4229-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 94.0 area%
  • Purity (with Total Nitrogen):
    min. 94.0 %

Specification Test Results

About this item

 :
 :1620.69 g/mol
Merck Index:14,02823
Storage Temperature:Freezer
CAS Number:103060-53-3
Formula:C₇₂H₁₀₁N₁₇O₂₆
MDL Number:MFCD08282794