Order Entry
Netherlands
ContactUsLinkComponent
2,2'-Dibromo-1,1'-binaphthyl ≥96.0% (by GC)
Irritant
2,2'-Dibromo-1,1'-binaphthyl ≥96.0% (by GC)
Supplier:  TCI
Certificates
Irritant
2,2'-Dibromo-1,1'-binaphthyl ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    182.0 to 186.0 deg-C

Specification Test Results

Frequently Bought Together