3-(1,3-Dithian-2-ylidene)-2,4-pentanedione ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:55727-23-6
:
Formula:C₉H₁₂O₂S₂
MDL Number:MFCD01096981
:216.31 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:101.0 to 105.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together